Ruthenium(III) bromide hydrate (≥25% Ru)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12628-03
12628-06
CAAA12628-03EA
115.34
CAD
CAAA12628-03
CAAA12628-06
Ruthenium(III) bromide hydrate (≥25% Ru)
Ruthenium(III) bromide hydrate
Uses: Oxidation
Soluble in alcohol
CAS No.: 14014-88-1
Molecular Formula: RuBr3.xH2O (x≈3)
Formula Weight: 340.78(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Odor: Characteristic
Solubility: Soluble in alcohol
UN No.: 3260
MDL No.: MFCD00016314
Applications: Oxidation
Formula:
RuBr₃·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00016314 CAS Number: 314075-07-5 EINECS: 237-829-4 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...