Palladium(II) nitrate (10% Pd) (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

12621-NB 12621-NH
CAAA12621-NBEA 1140.76 CAD
CAAA12621-NB CAAA12621-NH
Palladium(II) nitrate (10% Pd) (w/w) in aqueous solution
Palladium (II) nitrate

CAS No.: 10102-05-3 Molecular Formula: Pd(NO3)2 Formula Weight: 230.43 Physical Form: Liquid Appearance: Brown to yellow to orange UN No.: 3264 MDL No.: MFCD00011169

Formula: N₂O₆Pd
MW: 230.43
Storage Temperature: Ambient
MDL Number: MFCD00011169
CAS Number: 10102-05-3
EINECS: 233-265-8
UN: 3264
ADR: 8 II
Merck Index: 12,07124

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR