Niobium diboride ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals

12573-14 12573-22
CAAA12573-14EA 155.95 CAD
CAAA12573-14 CAAA12573-22
Niobium diboride ≥99% (metals basis)
Niobium diboride

Purity: 99% CAS No.: 12007-29-3 Molecular Formula: NbB2 Formula Weight: 114.53 Physical Form: Powder Odor: Odorless MDL No.: MFCD00016236

Formula: B₂Nb
Storage Temperature: Ambient
MDL Number: MFCD00016236
CAS Number: 12007-29-3
EINECS: 234-503-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR