Hafnium carbide ≥99.5% (metals basis excluding Zr) (max. 1% Zr)
Supplier: Thermo Scientific Chemicals
|
Warning
|
12567-09
12567-18
CAAA12567-03EA
41.63
CAD
CAAA12567-03
CAAA12567-09
CAAA12567-18
Hafnium carbide ≥99.5% (metals basis excluding Zr) (max. 1% Zr)
Hafnium carbide
Purity: 99.5%
CAS No.: 12069-85-1
Molecular Formula: CHf
Formula Weight: 190.54
Physical Form: -325 Mesh Powder
Density: 12.2
Appearance: Black
Odor: Odorless
Melting Point: 3890°
UN No.: 3178
MDL No.: MFCD00016126
Formula:
CHf Melting Pt: 3890 °C Density: 12.2 Storage Temperature: Ambient |
MDL Number:
MFCD00016126 CAS Number: 12069-85-1 EINECS: 235-114-1 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...