Chromium(III) acetate hydroxide (24% Cr)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chromium(III) acetate, basic

12518-36 12518-A3
CAAA12518-36EA 74.73 CAD
CAAA12518-36 CAAA12518-A3
Chromium(III) acetate hydroxide (24% Cr)
Chromium(III) acetate hydroxide

CAS No.: 39430-51-8 Molecular Formula: C14H23Cr3O16 Formula Weight: 603.31 Physical Form: Powder Appearance: Blue-green Odor: Odorless MDL No.: MFCD00058912

Formula: C₁₄H₂₃Cr₃O₁₆
MW: 603.311 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058912
CAS Number: 39430-51-8
EINECS: 254-447-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR