Nickel(II) hydroxide Ni 61%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12517-30
12517-A1
CAAA12517-30EA
184.86
CAD
CAAA12517-30
CAAA12517-A1
Nickel(II) hydroxide Ni 61%
Nickel (II) hydroxide
CAS No.: 12054-48-7
Molecular Formula: H2NiO2
Formula Weight: 92.70
Physical Form: Powder
Density: 4.1
Appearance: Green
Odor: Odorless
Melting Point: 230° dec.
UN No.: 3077
MDL No.: MFCD00011140
Formula:
H₂NiO₂ Melting Pt: 230 °C (dec.) Density: 4.1 Storage Temperature: Ambient |
MDL Number:
MFCD00011140 CAS Number: 12054-48-7 EINECS: 235-008-5 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...