Iron(II) chloride hydrate ≥99%, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Applications: Reducing agent in metallurgy, in pharmaceutical preparations, as mordant in dyeing and in sewage treatment
Purity: 99%
CAS No.: 23838-02-0
Molecular Formula: Cl2Fe.xH2O
Formula Weight: 126.75(anhy)
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Green-yellow
Odor: Odorless
Solubility: Soluble in water, alcohol and acetone. Insoluble in ether
UN No.: 3260
MDL No.: MFCD00149711
Applications: Reducing agent in metallurgy, in pharmaceutical preparations, as mordant in dyeing and in sewage treatment
Formula:
Cl₂Fe•xH₂O MW: 126.745 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00149711 CAS Number: 23838-02-0 EINECS: 231-843-4 UN: 3260 ADR: 8,III Merck Index: 14,4043 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...