Bismuth tin alloy (58:42 w%), eutectic ≥99.95% (metals basis), lumps
Supplier: Thermo Scientific Chemicals
12480-14
12480-22
12480-36
CAAA12480-14EA
71.1
CAD
CAAA12480-14
CAAA12480-22
CAAA12480-36
Bismuth tin alloy (58:42 w%), eutectic ≥99.95% (metals basis), lumps
Bismuth Tin alloy (58:42 w%), eutectic
Purity: 99.95%
Molecular Formula: Bi:Sn; 58:42 wt%
Melting Point: 138°
MDL No.: MFCD00143771
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...