Strontium zirconium oxide ≥99.3%

Supplier: Thermo Scientific Chemicals

Synonyms: Strontium zirconate

12346-30 12346-A1
CAAA12346-30EA 77.89 CAD
CAAA12346-30 CAAA12346-A1
Strontium zirconium oxide ≥99.3%
Strontium zirconium oxide

Purity: 99.3% CAS No.: 12036-39-4 Molecular Formula: SrZrO3 Formula Weight: 226.84 Physical Form: -325 Mesh Powder Density: 5.5 Appearance: White to off-white Odor: Odorless Melting Point: 2700° Boiling Point: 9000° MDL No.: MFCD00049555

Formula: SrZrO₃
MW: 226.841 g/mol
Boiling Pt: 9000 °C
Melting Pt: 2700 °C
Density: 5.5
Storage Temperature: Ambient
MDL Number: MFCD00049555
CAS Number: 12036-39-4
EINECS: 234-847-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR