Calcium zirconium oxide ≥99.2%

Supplier: Thermo Scientific Chemicals

12344-30 12344-A1
CAAA12344-30EA 70.94 CAD
CAAA12344-30 CAAA12344-A1
Calcium zirconium oxide ≥99.2%
Calcium zirconium oxide

Purity: 99.2% CAS No.: 12013-47-7 Molecular Formula: CaZrO3 Formula Weight: 179.30 Physical Form: -325 Mesh Powder Odor: Odorless Melting Point: 2550° MDL No.: MFCD00015982

Formula: CaZrO₃
MW: 179.30 g/mol
Melting Pt: 2550 °C
Storage Temperature: Ambient
MDL Number: MFCD00015982
CAS Number: 12013-47-7
EINECS: 234-586-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR