Molybdenum carbide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
12192-22
12192-36
CAAA12192-22EA
186.44
CAD
CAAA12192-22
CAAA12192-36
Molybdenum carbide ≥99.5% (metals basis)
Molybdenum carbide
Purity: 99.5%
CAS No.: 12069-89-5
Molecular Formula: Mo2C
Formula Weight: 203.91
Physical Form: -325 Mesh Powder
Odor: Odorless
MDL No.: MFCD00014218
Formula:
CMo₂ Melting Pt: 2687 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014218 CAS Number: 12069-89-5 EINECS: 235-115-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...