Dihydrogen hexachloroosmate(IV) hydrate (≥37% Os) ≥99.95% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chloroosmic acid, Hydrogen hexachloroosmate(IV)
12178-02
12178-03
CAAA12178-02EA
138.09
CAD
CAAA12178-02
CAAA12178-03
CAAA12178-06
Dihydrogen hexachloroosmate(IV) hydrate (≥37% Os) ≥99.95% (metals basis), Premion®
Dihydrogen hexachloroosmate(IV) hydrate
Purity: 99.95%
CAS No.: 27057-71-2
Molecular Formula: H2Cl6Os.xH2O
Formula Weight: 404.93(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Black
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD00011329
Formula:
H₂Cl₆Os.xH₂O MW: 404.93 g/mol (anhy) Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011329 CAS Number: 27057-71-2 EINECS: 248-192-7 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...