Ruthenium(III) nitrosylnitrate (≥31,3% Ru)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12175-03
12175-06
12175-14
CAAA12175-03EA
167.48
CAD
CAAA12175-03
CAAA12175-06
CAAA12175-14
Ruthenium(III) nitrosylnitrate (≥31,3% Ru)
Ruthenium(III) nitrosylnitrate
CAS No.: 34513-98-9
Molecular Formula: N4O10Ru
Formula Weight: 317.09
Physical Form: Crystalline
Appearance: Red-brown
Odor: Odorless
Melting Point: dec.
UN No.: 1477
MDL No.: MFCD00016313
Formula:
N₄O₁₀Ru MW: 317.09 g/mol Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00016313 CAS Number: 34513-98-9 EINECS: 252-068-8 UN: 1477 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...