Potassium pentachlororuthenate(III) hydrate (≥25.4% Ru) ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
12173-03
12173-06
CAAA12173-03EA
108.7
CAD
CAAA12173-03
CAAA12173-06
Potassium pentachlororuthenate(III) hydrate (≥25.4% Ru) ≥99.95% (metals basis)
Potassium pentachlororuthenate(III) hydrate
Purity: 99.95%
Molecular Formula: K2RuCl5.xH2O
Formula Weight: 356.54(anhy)
Physical Form: Crystalline
MDL No.: MFCD00058747
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...