Sodium tetrachloroplatinate(II) hydrate (≥42,4% Pt) ≥99.95%, Premion®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Disodium tetrachloroplatinate hydrate, Sodium platinum chloride hydrate, Sodium chloroplatinate(II) hydrate

12168-03 12168-06
CAAA12168-03EA 257.54 CAD
CAAA12168-03 CAAA12168-06
Sodium tetrachloroplatinate(II) hydrate (≥42,4% Pt) ≥99.95%, Premion®
Sodium tetrachloroplatinate(II) hydrate

Purity: 99.95% CAS No.: 207683-21-4 Molecular Formula: Na2PtCl4.xH2O (x≈3) Formula Weight: 382.88(anhy) Physical Form: Crystalline Melting Point: 100° MDL No.: MFCD00150188

Formula: Cl₄H₂Na₂OPt
MW: 382.88 g/mol (anhy)
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00150188
CAS Number: 207683-21-4
EINECS: 233-051-4

Order Now

Specification Test Results

Form Crystalline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR