Tetraammineplatinum(II) nitrate solution (3 - 4% Pt) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

12167-NA 12167-NJ
CAAA12167-NAEA 1104.42 CAD
CAAA12167-NA CAAA12167-NJ
Tetraammineplatinum(II) nitrate solution (3 - 4% Pt) (w/w)
Tetraammineplatinum(II) nitrate

CAS No.: 20634-12-2 Molecular Formula: H12N6O6Pt Formula Weight: 387.21 Physical Form: Liquid Density: 1.05 Appearance: Colorless UN No.: 3218 MDL No.: MFCD00011624

Formula: H₁₂N₆O₆Pt
MW: 387.216 g/mol
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00011624
CAS Number: 20634-12-2
EINECS: 243-929-9
UN: 3218
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR