Chromium disilicide 99+% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chromium silicide

12151-18 12151-30
CAAA12151-18EA 118.97 CAD
CAAA12151-18 CAAA12151-30
Chromium disilicide 99+% (metals basis)
Chromium disilicide

Purity: 99+% CAS No.: 12018-09-6 Molecular Formula: CrSi2 Formula Weight: 108.17 Physical Form: Powder Sensitivity: Moisture Sensitive MDL No.: MFCD00168070

Formula: CrSi₂
MW: 108.17 g/mol
Melting Pt: 1490 °C
Storage Temperature: Ambient
MDL Number: MFCD00168070
CAS Number: 12018-09-6
EINECS: 234-633-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR