Niobium carbide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

12147-14 12147-22
CAAA12147-14EA 87.06 CAD
CAAA12147-14 CAAA12147-22
Niobium carbide ≥99% (metals basis)
Niobium carbide

Purity: 99+% CAS No.: 12069-94-2 Molecular Formula: NbC Formula Weight: 104.91 Physical Form: 10 Micron Powder Density: 7.60 Melting Point: 3500° UN No.: 3178 MDL No.: MFCD00016237

Formula: CNb
Melting Pt: 3500 °C
Density: 7.60
Storage Temperature: Ambient
MDL Number: MFCD00016237
CAS Number: 12069-94-2
EINECS: 235-117-8
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR