Niobium nitride

Supplier: Thermo Scientific Chemicals

12146-09 12146-18
CAAA12146-09EA 119.92 CAD
CAAA12146-09 CAAA12146-18
Niobium nitride
Niobium nitride

CAS No.: 24621-21-4 Molecular Formula: NbN Formula Weight: 106.92 Physical Form: Powder Density: 8.40 Odor: Odorless MDL No.: MFCD00016239

Formula: NNb
MW: 106.92 g/mol
Melting Pt: 2573 °C
Density: 8.40
Storage Temperature: Ambient
MDL Number: MFCD00016239
CAS Number: 24621-21-4
EINECS: 246-362-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR