Zirconium diboride ≥99.5% (metals basis excluding Hf)

Supplier: Thermo Scientific Chemicals
Warning

12140-18 12140-30
CAAA12140-18EA 219.62 CAD
CAAA12140-18 CAAA12140-30
Zirconium diboride ≥99.5% (metals basis excluding Hf)
Zirconium diboride

Purity: 99.5% CAS No.: 12045-64-6 Molecular Formula: ZrB2 Formula Weight: 112.84 Physical Form: -325 Mesh Powder Melting Point: 3000° UN No.: 3178 MDL No.: MFCD00064648

Formula: B₂Zr
Melting Pt: 3000 °C
Storage Temperature: Ambient
MDL Number: MFCD00064648
CAS Number: 12045-64-6
EINECS: 234-963-5
UN: 3178
ADR: 4.1,III

Order Now

Specification Test Results

Form -325 Mesh Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR