Potassium perruthenate(VII) ≥97%

Supplier: Thermo Scientific Chemicals
Danger

11877-03 11877-06
CAAA11877-03EA 312.84 CAD
CAAA11877-03 CAAA11877-06
Potassium perruthenate(VII) ≥97%
Potassium perruthenate(VII)

Purity: 97% CAS No.: 10378-50-4 Molecular Formula: KO4Ru Formula Weight: 204.17 Physical Form: Crystalline Appearance: Black Melting Point: 440° dec. UN No.: 1479 MDL No.: MFCD00061333

Formula: C₃₂H₃₆N₂O₅
Melting Pt: 440 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00061333
CAS Number: 10378-50-4
EINECS: 233-835-6
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR