Potassium hexacyanoruthenate(II) hydrate (≥23,0% Ru)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium hexacyanoruthenate hydrate

11876-04 11876-09 11876-SS
CAAA11876-04EA 546.68 CAD
CAAA11876-04 CAAA11876-09 CAAA11876-SS
Potassium hexacyanoruthenate(II) hydrate (≥23,0% Ru)
Potassium hexacyanoruthenate(II) hydrate

CAS No.: 339268-21-2 Molecular Formula: C6K4N6Ru.xH2O Formula Weight: 413.57(anhy) Physical Form: Crystalline Appearance: White UN No.: 1588 MDL No.: MFCD00168064

Formula: C₆H₂K₄N₆ORu
Storage Temperature: Ambient
MDL Number: MFCD00168064
CAS Number: 339268-21-2
EINECS: 239-097-1
UN: 1588
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR