Indium(III) chloride hydrate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

11859-09 11859-18 11859-30
CAAA11859-09EA 142.67 CAD
CAAA11859-09 CAAA11859-18 CAAA11859-30
Indium(III) chloride hydrate ≥99.99% (metals basis)
Indium (III) chloride hydrate

Soluble in water


Purity: 99.99% CAS No.: 143983-91-9 Molecular Formula: InCl3.xH2O Formula Weight: 221.18(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Solubility: Soluble in water Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058

Formula: Cl₃In·H₂O
MW: 221.18 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149744
CAS Number: 143983-91-9
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 14,4958

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR