Indium(III) hydroxide ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
11855-09
11855-18
CAAA11855-09EA
155.16
CAD
CAAA11855-09
CAAA11855-18
Indium(III) hydroxide ≥99.998% (metals basis)
Indium (III) hydroxide
Purity: 99.998%
CAS No.: 20661-21-6
Molecular Formula: In(OH)3
Formula Weight: 165.84
Physical Form: -100 Mesh Powder
Melting Point: 150° -H2O
MDL No.: MFCD00011056
Formula:
H₃InO₃ Melting Pt: 150 °C (-H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00011056 CAS Number: 20661-21-6 EINECS: 243-947-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...