Ruthenium(III) iodide (≥20.5% Ru), anhydrous

Supplier: Thermo Scientific Chemicals
Warning

11805-03 11805-06
CAAA11805-03EA 154.21 CAD
CAAA11805-03 CAAA11805-06
Ruthenium(III) iodide (≥20.5% Ru), anhydrous
Ruthenium(III) iodide

CAS No.: 13896-65-6 Molecular Formula: RuI3 Formula Weight: 481.78 Physical Form: Powder Sensitivity: Hygroscopic Melting Point: 590° dec. MDL No.: MFCD00016316

Formula: I₃Ru
Melting Pt: 590 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016316
CAS Number: 13896-65-6
EINECS: 237-664-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR