Platinum 1% on gamma alumina, powder, reduced

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

11797-06 11797-14
CAAA11797-06EA 114.87 CAD
CAAA11797-06 CAAA11797-14 CAAA11797-22
Platinum 1% on gamma alumina, powder, reduced
Platinum

CAS No.: 7440-06-4 Molecular Formula: 300m2/g MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR