Ruthenium 10% on activated carbon, powder, reduced

Supplier: Thermo Scientific Chemicals
Danger

11757-14 11757-22
CAAA11757-14EA 469.26 CAD
CAAA11757-14 CAAA11757-22
Ruthenium 10% on activated carbon, powder, reduced
Ruthenium

Fieser: 1,983 5,574


CAS No.: 7440-18-8 Sensitivity: Air Sensitive Odor: Odorless UN No.: 1362 MDL No.: MFCD00011207

Formula: Ru
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 1362
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR