Ruthenium 5% on alumina, powder

Supplier: Thermo Scientific Chemicals

11749-06 11749-14 11749-22
CAAA11749-06EA 83.42 CAD
CAAA11749-06 CAAA11749-14 CAAA11749-22
Ruthenium 5% on alumina, powder
Ruthenium

Uses: Hydrogenation of aliphatic carbonyls and aromatic rings, especially bulky molecules


CAS No.: 7440-18-8 Physical Form: Powder Appearance: Greyish white Odor: Odorless MDL No.: MFCD00011207


Applications: Hydrogenation of aliphatic carbonyls and aromatic rings, especially bulky molecules

Formula: Ru
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR