Ruthenium 5% on activated carbon, powder, reduced

Supplier: Thermo Scientific Chemicals
Danger

11748-06 11748-14 11748-22
CAAA11748-06EA 81.21 CAD
CAAA11748-06 CAAA11748-14 CAAA11748-22
Ruthenium 5% on activated carbon, powder, reduced
Ruthenium

Aromatic ring hydrogenation


CAS No.: 7440-18-8 Odor: Odorless UN No.: 1362 MDL No.: MFCD00011207


Applications: Aromatic ring hydrogenation

Formula: Ru
MW: 101.07
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 1362
ADR: 4.2 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR