Palladium 5% on calcium carbonate, powder for use as a catalyst, reduced

Supplier: Thermo Scientific Chemicals

11723-06 11723-14 11723-22
CAAA11723-06EA 130.67 CAD
CAAA11723-06 CAAA11723-14 CAAA11723-22
Palladium 5% on calcium carbonate, powder for use as a catalyst, reduced
Palladium

CAS No.: 7440-05-3 Odor: Odorless MDL No.: MFCD03427452

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR