Palladium 5% on gamma alumina, powder, reduced

Supplier: Thermo Scientific Chemicals

11713-06 11713-14 11713-22
CAAA11713-06EA 130.67 CAD
CAAA11713-06 CAAA11713-14 CAAA11713-22
Palladium 5% on gamma alumina, powder, reduced
Palladium

CAS No.: 7440-05-3 Physical Form: Powder Odor: Odorless MDL No.: MFCD03613602

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03613602
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR