Sodium hexanitritocobaltate(III) ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
11667-22
11667-36
CAAA11667-22EA
206.98
CAD
CAAA11667-22
CAAA11667-36
Sodium hexanitritocobaltate(III) ACS
Sodium hexanitritocobaltate(III)
Gradual decomposition of aqueous solutions is slowed by small amounts of acetic acid
Very soluble in water
CAS No.: 14649-73-1
Molecular Formula: Na3Co(NO2)6
Formula Weight: 403.94
Physical Form: Powder
Appearance: Yellow to orange
Odor: Odorless
Melting Point: 220° dec.
Solubility: Very soluble in water
UN No.: 1479
MDL No.: MFCD00003512
Formula:
Na₃Co(NO₂)₆ Melting Pt: 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003512 CAS Number: 13600-98-1 EINECS: 237-077-7 UN: 1479 ADR: 5.1,II Merck Index: 14,8603 |
Specification Test Results
Insoluble matter | 0.02% max. |
Suitability for determination of potassium | Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...