Cerium (III) perchlorate hexahydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

11654-18 11654-30
CAAA11654-18EA 88.95 CAD
CAAA11654-18 CAAA11654-30
Cerium (III) perchlorate hexahydrate, Reagent Grade
Cerium (III) perchlorate hexahydrate

CAS No.: 14017-47-1 Molecular Formula: Ce(ClO4)3.6H2O Formula Weight: 546.56 (438.47anhy) Physical Form: Crystalline Sensitivity: Moisture Sensitive Odor: Odorless UN No.: 1481 MDL No.: MFCD00150205

Formula: CeCl₃O₁₂·6H₂O
MW: 546.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150205
CAS Number: 14017-47-1
EINECS: 237-836-2
UN: 1481
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR