Iron(III) perchlorate hydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

11647-22 11647-36
CAAA11647-22EA 97.01 CAD
CAAA11647-22 CAAA11647-36
Iron(III) perchlorate hydrate, Reagent Grade
Iron(III) perchlorate hydrate

CAS No.: 15201-61-3 Molecular Formula: Fe(ClO4)3.xH2O Formula Weight: 354.19(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Yellow UN No.: 1481 MDL No.: MFCD00011000

Formula: Cl₃FeH₂O₁₃
MW: 372.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011000
CAS Number: 15201-61-3
EINECS: 236-908-0
UN: 1481
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR