Iron(II) perchlorate hydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals

11646-22 11646-36
CAAA11646-22EA 81.69 CAD
CAAA11646-22 CAAA11646-36
Iron(II) perchlorate hydrate, Reagent Grade
Iron(II) perchlorate hydrate

CAS No.: 335159-18-7 Molecular Formula: Fe(ClO4)2.xH2O Formula Weight: 254.75(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 100° dec. UN No.: 1481 MDL No.: MFCD00150234

Formula: Fe(ClO₄)₂.xH₂O
MDL Number: MFCD00150234
CAS Number: 335159-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR