Molybdenum disilicide Mo ≥ 61% , ∼99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Molybdenum silicide
Applications: In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles
Insoluble in acids and aqua regia. Very soluble in HF and HNO3
Purity: 99.5%
CAS No.: 12136-78-6
Molecular Formula: MoSi2
Formula Weight: 152.13
Physical Form: -325 Mesh Powder
Density: 6.31
Melting Point: 1870-2030°
Solubility: Insoluble in acids and aqua regia, however, it is soluble in HF and HNO3
MDL No.: MFCD00151535
In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles
Formula:
MoSi₂ Melting Pt: 1870…2030 °C Density: 6,31 Storage Temperature: Ambient |
MDL Number:
MFCD00151535 CAS Number: 12136-78-6 EINECS: 235-231-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...