Trichromium dicarbide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
11547-18
11547-30
CAAA11547-18EA
114.55
CAD
CAAA11547-18
CAAA11547-30
Trichromium dicarbide ≥99.5% (metals basis)
Trichromium dicarbide
Purity: 99.5%
CAS No.: 12012-35-0
Molecular Formula: Cr3C2
Formula Weight: 180.02
Physical Form: -325 Mesh Powder
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00016041
Formula:
C₂Cr₃ Melting Pt: 1890 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016041 CAS Number: 12012-35-0 EINECS: 234-576-1 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...