Silver phosphate ≥99%

Supplier: Thermo Scientific Chemicals
Danger

11415-06 11415-14
CAAA11415-06EA 44.08 CAD
CAAA11415-06 CAAA11415-14
Silver phosphate ≥99%
Silver phosphate

Applications: In photography
Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3, ammonia, ammonium carbonate, alkali cyanides and thiosulfates


Purity: 99% CAS No.: 7784-09-0 Molecular Formula: Ag3O4P Formula Weight: 418.58 Physical Form: Powder Density: 6.37 Sensitivity: Light Sensitive Appearance: Yellow Odor: Odorless Melting Point: 849° Solubility: Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3, ammonia, ammonium carbonate, alkali cyanides and thiosulfates UN No.: 3077 MDL No.: MFCD00014148


Applications: In photography

Formula: Ag₃O₄P
MW: 418.58 g/mol
Melting Pt: 849 °C
Density: 6.37
Storage Temperature: Ambient
MDL Number: MFCD00014148
CAS Number: 7784-09-0
EINECS: 232-049-0
UN: 3077
ADR: 9 III
Merck Index: 14,8525

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR