Cerium(IV) sulfate hydrate ≥99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11323-22
11323-36
CAAA11323-22EA
170.64
CAD
CAAA11323-22
CAAA11323-36
Cerium(IV) sulfate hydrate ≥99% (REO, rare earth oxide basis), REacton®
Cerium (IV) sulphate hydrate
Fieser: 21,94
Purity: 99%
CAS No.: 95838-16-7
Molecular Formula: Ce(SO4)2.xH2O
Formula Weight: 332.24(anhy)
Physical Form: Crystalline
Density: 3.01
Melting Point: 195° dec.
UN No.: 3085
MDL No.: MFCD00149427
Formula:
CeO₈S₂·H₂O MW: 332.24 g/mol (anhy) Melting Pt: 195 °C (dec.) Density: 3,01 Storage Temperature: Ambient |
MDL Number:
MFCD00149427 CAS Number: 95838-16-7 EINECS: 237-029-5 UN: 3085 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...