Holmium(III) chloride, hydrate ≥99.99%, REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Holmium trichloride

11276-09 11276-18
CAAA11276-09EA 241.74 CAD
CAAA11276-09 CAAA11276-18
Holmium(III) chloride, hydrate ≥99.99%, REacton®
Holmium(III) chloride

Purity: 99.99% Molecular Formula: HoCl3.xH2O Formula Weight: 271.29(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Melting Point: 718° MDL No.: MFCD00011051

Formula: Cl₃Ho
MW: 271.29
Boiling Pt: 1500
Melting Pt: 718 °C
Density: 3.7
Storage Temperature: Ambient
MDL Number: MFCD00011051
CAS Number: 10138-62-2
EINECS: 233-387-1
Merck Index: 14,4727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR