Lanthanum triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
Synonyms:
Lanthanum(III) acetate sesquihydrate
11263-18
11263-30
CAAA11263-18EA
172.22
CAD
CAAA11263-18
CAAA11263-30
Lanthanum triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum triacetate sesquihydrate
Purity: 99.99%
CAS No.: 25721-92-0
Molecular Formula: C6H9LaO6.1.5H2O
Formula Weight: 343.05 (316.03anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00013037
Formula:
C₆H₉LaO₆·1,5H₂O MW: 343.05 g/mol (316.03 anhy) Melting Pt: <200 °C (-1.5H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00013037 CAS Number: 25721-92-0 EINECS: 213-034-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...