Neodymium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11245-06
11245-14
CAAA11245-06EA
73.31
CAD
CAAA11245-06
CAAA11245-14
Neodymium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Neodymium trinitrate hydrate
Purity: 99.99%
CAS No.: 13746-96-8
Molecular Formula: Nd(NO3)3.xH2O
Formula Weight: 330.25(anhy)
Physical Form: Wet Crystalline Aggregates
Sensitivity: Hygroscopic
pH: 3.0-4.0
Melting Point: ca 40°
Merck Reference: 14,6450
UN No.: 1477
MDL No.: MFCD00149798
Formula:
N₃NdO₉·H₂O Melting Pt: ∼ 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149798 CAS Number: 13746-96-8 EINECS: 233-153-9 UN: 1477 ADR: 5.1,III Merck Index: 14,6450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...