Yttrium(III) acetate tetrahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
11186-14
11186-22
CAAA11186-14EA
77.58
CAD
CAAA11186-14
CAAA11186-22
Yttrium(III) acetate tetrahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium(III) acetate tetrahydrate
Purity: 99.99%
CAS No.: 85949-60-6
Molecular Formula: C6H9O6Y.4H2O
Formula Weight: 338.10 (266.04anhy)
Physical Form: Crystalline Aggregates
Density: 1.5
Appearance: White
Melting Point: dec.
MDL No.: MFCD00013048
Formula:
C₆H₉O₆Y·4H₂O MW: 338.10 g/mol (266.04 anhy) Melting Pt: Dec. Density: 1.5 Storage Temperature: Ambient |
MDL Number:
MFCD00013048 CAS Number: 85949-60-6 EINECS: 245-612-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...