Gallium(III) sulfate hydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
11152-09
11152-18
CAAA11152-09EA
308.1
CAD
CAAA11152-09
CAAA11152-18
Gallium(III) sulfate hydrate ≥99.999% (metals basis), Puratronic®
Gallium(III) sulfate hydrate
Purity: 99.999%
CAS No.: 13780-42-2
Molecular Formula: Ga2(SO4)3.xH2O (x≈18)
Formula Weight: 427.62(anhy)
Physical Form: Crystalline Aggregates
Density: 3.860
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 105-110°
MDL No.: MFCD00149725
Formula:
Ga2(SO4)3•xH2O (x≈18) Melting Pt: 105…110 °C Density: 3.860 Storage Temperature: Ambient |
MDL Number:
MFCD00149725 CAS Number: 13780-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...