Dihydrogen hexahydroxyplatinate(IV) (≥61,0% Pt) ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
11052-02
11052-03
11052-06
CAAA11052-02EA
161.16
CAD
CAAA11052-02
CAAA11052-03
CAAA11052-06
Dihydrogen hexahydroxyplatinate(IV) (≥61,0% Pt) ≥99.9% (metals basis)
Dihydrogen hexahydroxyplatinate(IV)
Purity: 99.9%
CAS No.: 51850-20-5
Molecular Formula: H2Pt(OH)6
Formula Weight: 299.17
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Pale to dark yellow
Melting Point: dec.
MDL No.: MFCD00058744
Formula:
H₂Pt(OH)₆ Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00058744 CAS Number: 51850-20-5 EINECS: 257-471-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...