trans-Diamminedichloropalladium(II) (≥49.9% Pd) ≥99.95% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trans-Dichlorodiammine palladium(II), Dichlorodiamminepalladium, trans-Diammine dichloropalladium

11037-02 11037-09
CAAA11037-02EA 140.94 CAD
CAAA11037-02 CAAA11037-09
trans-Diamminedichloropalladium(II) (≥49.9% Pd) ≥99.95% (metals basis), Premion®
trans-Diamminedichloropalladium(II)

Slightly soluble in water. Soluble in NH3OH. Insoluble in chloroform, acetone


Purity: 99.95% CAS No.: 14323-43-4 Molecular Formula: Pd(NH3)2Cl2 Formula Weight: 211.37 Physical Form: Crystalline powder Density: 2.5 Appearance: Yellow Solubility: Slightly soluble in water. Soluble in NH4OH. Insoluble in chloroform, acetone UN No.: 3077 MDL No.: MFCD00011621

Formula: Pd(NH₃)₂Cl₂
MW: 211.39 g/mol
Density: 2.5
Storage Temperature: Ambient
MDL Number: MFCD00011621
CAS Number: 14323-43-4
EINECS: 238-269-3
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR