Tetraamminepalladium(II) chloride monohydrate (≥39% Pd) ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
11036-04
11036-09
CAAA11036-04EA
448.72
CAD
CAAA11036-04
CAAA11036-09
Tetraamminepalladium(II) chloride monohydrate (≥39% Pd) ≥99.9% (metals basis)
Tetraamminepalladium(II) chloride monohydrate
Purity: 99.9%
CAS No.: 13933-31-8
Molecular Formula: Pd(NH3)4Cl2.H2O
Formula Weight: 263.45 (245.43anhy)
Physical Form: Crystalline
Density: 1.91
Sensitivity: Hygroscopic
Appearance: Colorless to yellow
Melting Point: 120° dec.
UN No.: 3077
MDL No.: MFCD00151033
Formula:
Cl₂H₁₄N₄OPd Melting Pt: 120 °C (dec.) Density: 1,91 Storage Temperature: Ambient |
MDL Number:
MFCD00151033 CAS Number: 13933-31-8 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...