Ammonium bis(oxalato)oxotitanate(IV) hydrate ≥99.998%, Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10895-09 10895-18
CAAA10895-03EA 136.54 CAD
CAAA10895-03 CAAA10895-09 CAAA10895-18
Ammonium bis(oxalato)oxotitanate(IV) hydrate ≥99.998%, Puratronic®
Ammonium bis(oxalato)oxotitanate(IV) hydrate

Purity: 99.998% CAS No.: 10580-03-7 Molecular Formula: (NH4)2TiO(C2O4)2.xH2O (x≈1) Formula Weight: 276.01(anhy) Physical Form: Crystalline powder Odor: Odorless UN No.: 3288 MDL No.: MFCD00149620

Formula: (NH₄)₂TiO(C₂O₄)₂·xH₂O (x≈1)
MW: 276.01 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149620
CAS Number: 10580-03-7
EINECS: 234-174-6
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR