Tetraammineplatinum(II) chloride monohydrate ≥98%, Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetraammineplatinum (II) chloride hydrate
10836-03
10836-06
CAAA10836-03EA
328.64
CAD
CAAA10836-03
CAAA10836-06
Tetraammineplatinum(II) chloride monohydrate ≥98%, Premion®
Tetraammineplatinum(II) chloride monohydrate
Purity: 99.995%
CAS No.: 13933-33-0
Molecular Formula: Pt(NH3)4Cl2.H2O
Formula Weight: 352.13 (334.11anhy)
Physical Form: Crystalline
Density: 2.74
Sensitivity: Hygroscopic
Appearance: White to cream
Melting Point: 250°dec.
MDL No.: MFCD00149947
Formula:
Cl₂H₁₄N₄OPt MW: 352.123 g/mol Melting Pt: 250 °C. (dec.) Density: 2.74 Storage Temperature: Ambient |
MDL Number:
MFCD00149947 CAS Number: 13933-33-0 EINECS: 237-706-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...