Nickel sulfate hydrate ≥99.9985% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10820-14
10820-22
CAAA10820-14EA
153.42
CAD
CAAA10820-14
CAAA10820-22
Nickel sulfate hydrate ≥99.9985% (metals basis), Puratronic®
Nickel sulphate hydrate
m.p. 100° -5H2O, 280° -6H2O
Purity: 99.9985%
CAS No.: 15244-37-8
Molecular Formula: NiO4S.xH2O (x≈6)
Formula Weight: 154.77(anhy)
Physical Form: Crystalline powder
Density: 2.070
Appearance: Green
Odor: Odorless
pH: 4.3-4.7
Merck Reference: 14,6517
UN No.: 3288
MDL No.: MFCD00011146
Formula:
NiO₄S·H₂O Density: 2.070 Storage Temperature: Ambient |
MDL Number:
MFCD00011146 CAS Number: 15244-37-8 EINECS: 232-104-9 UN: 3288 ADR: 6.1,III Merck Index: 14,6517 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...